1-Hexadecanethiol 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2917-26-2
:
Formula:C₁₆H₃₄S
MDL Number:MFCD00011677
:258.51 g/mol
Boiling Pt:184…191 °C (7 mmHg)
Melting Pt:18…20 °C
Density:0.84 g/cm³
Flash Pt:101 °C
Storage Temperature:Ambient
EINECS:220-846-6
ADR:9,
Specification Test Results
- Appearance:Slightly hazy colorless to white to light yellow Liquid or waxy solid
- Infrared spectrum:Conforms
- Refractive index:1.4610 to 1.4640 (20°C, 589 nm)
- GC:≥88 %
- Stabilizer:Stabilized with BHT
Specification Test Results
Frequently Bought Together