
Clelandovo činidlo, bílý, solid
Catalog # 91050.
Supplier: Biotium
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
Boiling Pt:∼125 °C (1013 hPa)
Melting Pt:42…44 °C
Flash Pt:>110 °C
EINECS:222-468-7
:GHS07
Storage Temperature:Refrigerator
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877