Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Dithiothreitol (DTT, Cleland's reagent) 99% for biochemistry
 
undefined
 
Dithiothreitol (DTT, Cleland's reagent) 99% for biochemistry

 

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    38 to 43 °C
  • Titration Iodimetric:
    ≥98.5 %
  • UV:
    (1 M in water)
  • Appearance of solution:
    (5 % in water) Clear colorless
  • UV transmittance:
    ≥99 % (c=5, 660 nm, water) 1 cm cell

 

 

Frequently Bought Together