Order Entry
Czech Republic
ContactUsLinkComponent
2,3-Dimercapto-1-propanol ≥98.0% (by GC) for complexometry, for spectrophotometric determination of Cu, Fluka™
Toxic
2,3-Dimercapto-1-propanol ≥98.0% (by GC) for complexometry, for spectrophotometric determination of Cu, Fluka™
Catalog # 38520-25ML
CAS Number:  
undefined
Toxic
2,3-Dimercapto-1-propanol ≥98.0% (by GC) for complexometry, for spectrophotometric determination of Cu, Fluka™
Catalog # 38520-25ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Refractive Index:
    n20/D 1.573 (lit.)
  • Assay (GC):
    Min. 98.0 Fl.%
  • IR spectrum:
    complying
  • Refractive index (n 20/D):
    1.573 - 1.575

Specification Test Results

About this item

 :
Merck Index:13,03234
Boiling Pt:120 °C (20 hPa)
Melting Pt:77 °C
Density:1.24 g/cm³ (25 °C)
Flash Pt:112 °C
Storage Temperature:Ambient
EINECS:200-433-7
UN:2810
ADR:6.1,III
 :124.22 g/mol
 :GHS06
CAS Number:59-52-9
Formula:C₃H₈OS₂
MDL Number:MFCD00004864