Order Entry
Czech Republic
ContactUsLinkComponent
1-Tetradecanol
1-Tetradecanol
Catalog # EHERC17396800
Supplier:  EHRENSTORFER
CAS Number:  
1-Tetradecanol
Catalog # EHERC17396800
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :214.39 g/mol
Merck Index:13,06361
Boiling Pt:289 °C (1013 hPa)
Melting Pt:37…39 °C
Density:0.8 g/cm³ (60 °C)
Storage Temperature:Ambient
EINECS:204-000-3
CAS Number:112-72-1
Formula:H₃C(CH₂)₁₂CH₂OH
MDL Number:MFCD00004757