Order Entry
Czech Republic
ContactUsLinkComponent
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Irritant
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # 293245-1L
CAS Number:  
undefined
Irritant
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # 293245-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Non-volatile matter:
    Max. 0.004 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 300 nm:
    Max. 0.01
  • Absorbance at 250 nm:
    Max. 0.05
  • Absorbance at 235 nm:
    Max. 0.20
  • Absorbance at 225 nm:
    Max. 0.50
  • Absorbance at 215 nm:
    Max. 1.0
  • APHA:
    Max. 10
  • Refractive Index:
    n20/D 1.429 (lit.)

Specification Test Results

About this item

 :
 :130.23 g/mol
Merck Index:13,06784
Boiling Pt:194 °C (1.013 hPa)
Melting Pt:-15 °C
Melting Pt:–15 °C
Density:0.827 g/cm³ (25 °C)
Flash Pt:80 °C
Storage Temperature:Ambient
EINECS:203-917-6
 :GHS07
CAS Number:111-87-5
Formula:H₃C(CH₂)₆CH₂OH
MDL Number:MFCD00002988