
1-Octanol 99%, Reagent Grade
Catalog # 112615-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Refractive index (n 20/D):1.428 - 1.431
- Non-volatile matter:Max. 0.1 %
- Water (Karl Fischer):Max. 0.5 %
- Free acid (as CH₃COOH):Max. 0.01 %
- Chloride (Cl):complying
- APHA:Max. 20
Specification Test Results
About this item
:
:130.23 g/mol
Merck Index:13,06784
Boiling Pt:194 °C (1.013 hPa)
Melting Pt:-15 °C
Melting Pt:–15 °C
Density:0.827 g/cm³ (25 °C)
Flash Pt:80 °C
Storage Temperature:Ambient
EINECS:203-917-6
:GHS07
CAS Number:111-87-5
Formula:H₃C(CH₂)₆CH₂OH
MDL Number:MFCD00002988