

Bis(2-methoxyethyl)ether, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-96-6
:
Formula:C₆H₁₄O₃
MDL Number:MFCD00008503
Merck Index:13,03187
Boiling Pt:162 °C (1013 hPa)
Melting Pt:–64 °C
Density:0.944 g/cm³ (20 °C)
Flash Pt:53 °C
EINECS:203-924-4
UN:3271
ADR:3,III
:M-Clarity™ quality level = MQ100
:134.18 g/mol
:GHS02
:GHS08
Storage Temperature:Ambient
Frequently Bought Together