Order Entry
Czech Republic
ContactUsLinkComponent
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3% stabilised ACS, Riedel-de Haën™
Flammable
Health
Irritant
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3% stabilised ACS, Riedel-de Haën™
Catalog # SIAL360503-1L
Supplier:  Merck
CAS Number:  
Flammable
Health
Irritant
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3% stabilised ACS, Riedel-de Haën™
Catalog # SIAL360503-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    99.3%

Specification Test Results

About this item

 :
 :76.1 g/mol
Merck Index:12,06118
Boiling Pt:124.6 °C (1013 hPa)
Melting Pt:–85 °C
Density:0.964 g/cm³ (20 °C)
Flash Pt:39 °C
Storage Temperature:Ambient
EINECS:203-713-7
UN:1188
ADR:3,III
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 50 ppm
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-86-4
Formula:C₃H₈O₂
MDL Number:MFCD00002867