


N-Ethyldiisopropylamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7087-68-5
:
Formula:C₈H₁₉N
MDL Number:MFCD00008868
:129.25 g/mol
Boiling Pt:127 °C (1013 hPa)
Melting Pt:–50 °C
Density:0.76 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
UN:2733
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:230-392-0
:GHS02
:GHS06
:GHS05
Frequently Bought Together