


N-Ethyldiisopropylamine ≥99.5%, Biotechnology Grade, Burdick & Jackson™
Catalog # 496219-2L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 L
- Seq:5086
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.45 %
- Water (Karl Fischer):Max. 0.05 %
- APHA:Max. 20
- Vapor Pressure:31 mmHg ( 37.7 °C)
Specification Test Results
About this item
:
:129.25 g/mol
Boiling Pt:128.33 °C
Melting Pt:-19.99 °C
Melting Pt:–19.99 °C
Density:0.75 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:230-392-0
UN:2733
ADR:3,II
:GHS05
:GHS06
:GHS02
CAS Number:7087-68-5
Formula:C₈H₁₉N
MDL Number:MFCD00008868