


N-Ethyldiisopropylamine 99.5+%
:7087-68-5
:
:C₈H₁₉N
:MFCD00008868
:129.25 g/mol
:127 °C (760 mmHg)
:–46 °C
:0.756 g/cm³
:6 °C
:Ambient
:230-392-0
:3384
:6.1,
:GHS02
:GHS05
:GHS06
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4130 to 1.4150 (20°C, 589 nm)
- Water:≤0.3 %
- GC:≥99.50 %
- Color scale:≤50 APHA
Frequently Bought Together