


Diisopropylamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-18-9
:
Formula:C₆H₁₅N
MDL Number:MFCD00008862
:101.19 g/mol
Merck Index:13,03219
Boiling Pt:84 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.717 g/cm³ (20 °C)
EINECS:203-558-5
UN:1158
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:–17 °C
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together