Order Entry
Czech Republic
ContactUsLinkComponent
Diisopropylamine ≥99.0% for synthesis, Sigma-Aldrich®
Flammable
Toxic
Corrosive
Diisopropylamine ≥99.0% for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Toxic
Corrosive
Diisopropylamine ≥99.0% for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (GC area %):
    ≥ 99.0 % (a/a)
  • Density (d 20 °C/ 4 °C):
    0.715 - 0.716
  • Water (K. F.):
    ≤ 0.30 %
  • Identity (IR):
    passes test

Specification Test Results

Related Information

Frequently Bought Together