


Diisopropylamine 99.5%, AcroSeal®, redistilled
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-18-9
:
Formula:C₆H₁₅N
MDL Number:MFCD00008862
:101.19 g/mol
Merck Index:13,03219
Boiling Pt:84 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.716 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
EINECS:203-558-5
UN:1158
ADR:3,II
:GHS06
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3910 to 1.3930 (20°C, 589 nm)
- GC:≥99.4 %
- Color scale:≤10 APHA
Specification Test Results
Related Information
Frequently Bought Together