


1,2-Propylenediamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-90-0
:
Formula:C₃H₁₀N₂
MDL Number:MFCD00008089
:74.13 g/mol
Merck Index:14,7852
Boiling Pt:119…120 °C
Melting Pt:∼–37 °C
Density:0.869
Flash Pt:34 °C (93 °F)
Storage Temperature:Ambient
EINECS:201-155-9
UN:2258
ADR:8,II
:GHS05
:GHS02
:GHS07
Related Information
Frequently Bought Together