

Ethanolamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:141-43-5
:
Formula:NH₂CH₂CH₂OH
MDL Number:MFCD00008183
Merck Index:13,03762
Boiling Pt:171 °C (1013 hPa)
Melting Pt:10.5 °C
Density:1.0164 g/cm³ (20 °C)
Flash Pt:93 °C
EINECS:205-483-3
UN:2491
ADR:8,III
:M-Clarity™ quality level = MQ200
:61.08 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:10.5 °C
- Molar Mass:61.08 g/mol
- Boiling Point:171 °C (1013 hPa)
- Vapor Pressure:0.5 hPa (20 °C)
- Flash Point:92.5 °C
- Refractive Index:1.4539
- Explosion Limit:3.4 - 27 %(V)
- Density:1.02 g/cm3 (20 °C)
- pH:12.1 (100 g/l, H2O, 20 °C)
- Ignition Point:410 °C
- Viscosity kinematic:20 mm2/s (23 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
Related Information
Frequently Bought Together