Order Entry
Czech Republic
ContactUsLinkComponent
Ethanolamine, (max. 0.5% H₂O) ≥99%
Ethanolamine, (max. 0.5% H₂O) ≥99%
  22793.30
 :  
undefined
Ethanolamine, (max. 0.5% H₂O) ≥99%
  22793.30
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545
  • Environmentally Preferable:

 

 

 : 2-Aminoethanol, Monoethanolamine
 :
 :61.08 g/mol
 :10 °C
 :205-483-3
 :2491
 :GHS05
 :GHS07
 :Ambient
 :141-43-5
 :NH₂CH₂CH₂OH
 :MFCD00008183
 :169…170 °C
 :1.012
 :93 °C (199 °F)
 :8 III
 :14,3727