

1,6-Diisocyanatohexane for synthesis, Sigma-Aldrich®
: Merck
:822-06-0
:
:C₈H₁₂N₂O₂
:MFCD00002047
:168.2 g/mol
:255 °C (1013 hPa)
:–67 °C
:1.05 g/cm³ (20 °C)
:130 °C
:2281
:6.1,II
:M-Clarity™ quality level = MQ200
:212-485-8
:GHS06
:GHS08
:Ambient
- Solubility:(20 °C) (decomposition)
- Melting Point:-67 °C
- Molar Mass:168.2 g/mol
- Boiling Point:255 °C (1013 hPa)
- Vapor Pressure:0.007 hPa (20 °C)
- Flash Point:130 °C
- Refractive Index:1.4525 (20 °C, 589 nm)
- Explosion Limit:0.9 - 9.5 %(V)
- Density:1.05 g/cm3 (25 °C)
- Ignition Point:454 °C
- Synonyms:1,6-Diisocyanatohexane
- Hill Formula:C8H12N2O2
- Chemical Formula:OCN(CH2)6NCO
Frequently Bought Together