Order Entry
Czech Republic
ContactUsLinkComponent
1,2-Bis(dimethylamino)methane 99%
Flammable
1,2-Bis(dimethylamino)methane 99%
Certificates
undefined
Flammable
1,2-Bis(dimethylamino)methane 99%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3995 to 1.4025 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together