


N,N,N',N'-Tetramethylethylenediamine (TEMED), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-18-9
:
Formula:C₆H₁₆N₂
MDL Number:MFCD00008335
:116.21 g/mol
Boiling Pt:120 °C (1013 hPa)
Melting Pt:–64 °C
Density:0.7765 g/cm³ (20 °C)
Flash Pt:17 °C (closed cup)
Storage Temperature:Ambient
UN:2372
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:203-744-6
:GHS02
:GHS05
:GHS07
Frequently Bought Together