


N,N-Dimethylethylendiamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-00-9
:
Formula:C₄H₁₂N₂
MDL Number:MFCD00008175
:88.15 g/mol
Boiling Pt:∼106 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.804 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:12 °C
EINECS:203-541-2
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together