Order Entry
Czech Republic
ContactUsLinkComponent
N,N-Dimethylethylendiamine for synthesis, Sigma-Aldrich®
 
 
 
N,N-Dimethylethylendiamine for synthesis, Sigma-Aldrich®
  8.03779.0100
 :  Merck
 :  
 
 
 
N,N-Dimethylethylendiamine for synthesis, Sigma-Aldrich®
  8.03779.0100
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    88.15 g/mol
  • Boiling Point:
    107 - 109 °C (1013 hPa)
  • Vapor Pressure:
    29 hPa (20 °C)
  • Flash Point:
    12.5 °C
  • Explosion Limit:
    1.3 - 10.8 %(V)
  • Density:
    0.81 g/cm3 (20 °C)
  • pH:
    11.4 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    225 °C

 

 

 :
 :88.15 g/mol
 :∼106 °C (1013 hPa)
 :–70 °C
 :0.804 g/cm³ (20 °C)
 :12 °C
 :Ambient
 :203-541-2
 :2733
 :3 (8),II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS05
 :GHS07
 :108-00-9
 :C₄H₁₂N₂
 :MFCD00008175