Order Entry
Czech Republic
ContactUsLinkComponent
Triethylsilyl trifluoromethanesulphonate 99%
Corrosive
Triethylsilyl trifluoromethanesulphonate 99%
Certificates
undefined
Corrosive
Triethylsilyl trifluoromethanesulphonate 99%

Specification Test Results

  • Appearance:
    Clear colorless to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3890 to 1.3920 (20°C, 589 nm)
  • Titration excess NaOH:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together