Order Entry
Czech Republic
ContactUsLinkComponent
Trifluoromethanesulphonyl chloride 99%
Corrosive
Trifluoromethanesulphonyl chloride 99%
Certificates
undefined
Corrosive
Trifluoromethanesulphonyl chloride 99%

Specification Test Results

  • Appearance:
    Clear colorless to slightly colored Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3330 to 1.3350 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together