2,2,3,3-Tetrafluoro-1-propanol, Sigma-Aldrich®
Catalog # SIAL326275-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:132.06 g/mol
Merck Index:12,09346
Boiling Pt:109 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.6647 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-955-5
UN:1987
ADR:3,III
:M-Clarity™ quality level = MQ200
CAS Number:76-37-9
Formula:C₃H₄F₄O
MDL Number:MFCD00004676