
Pentafluoropropionic anhydride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:356-42-3
:
Formula:C₆F₁₀O₃
MDL Number:MFCD00000429
:310.05 g/mol
Boiling Pt:69 °C (1013 hPa)
Density:1.571 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:206-604-2
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
Frequently Bought Together