Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane, Sigma-Aldrich®
Catalog # SIAL370525-25G
Supplier:  Merck
CAS Number:  
Irritant
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane, Sigma-Aldrich®
Catalog # SIAL370525-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :574.02 g/mol
Boiling Pt:122 °C (50 mmHg)
Melting Pt:55.5…56.5 °C
Density:1.88 g/cm³ (60 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:218-053-5
 :GHS07
CAS Number:2043-53-0
Formula:C₁₀H₄F₁₇I
MDL Number:MFCD00039411