Order Entry
Czech Republic
ContactUsLinkComponent
EDTA (ethylenediamine tetraacetic acid) 98.0-100.5%, GPR RECTAPUR®
Irritant
EDTA (ethylenediamine tetraacetic acid) 98.0-100.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
EDTA (ethylenediamine tetraacetic acid) 98.0-100.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    98.0 - 100.5 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Water:
    Max. 0.5 %
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

Frequently Bought Together