
Triammonium citrate 97-103%, GPR RECTAPUR®
Catalog # 271534H
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay:97 - 103 %
- Identification:Passes test
- pH (20°C; 5 %):6.0 - 7.5
- Ignition residue (SO4):Max. 0.05 %
- Cl (Chloride):Max. 100 ppm
- Pb (Lead):Max. 10 ppm
Specification Test Results
About this item
:
:243.22 g/mol
Melting Pt:185 °C
Density:1 g/cm³ (25 °C)
EINECS:222-394-5
:GHS07
Storage Temperature:Ambient
CAS Number:3458-72-8
Formula:C₆H₁₇N₃O₇
MDL Number:MFCD00036406