Order Entry
Czech Republic
ContactUsLinkComponent
Potassium citrate monohydrate, granular USP, FCC, J.T.Baker®
Potassium citrate monohydrate, granular USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Potassium citrate monohydrate, granular USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Identification:
    Passes Test
  • USP - Alkalinity:
    Passes Test
  • USP - Loss on Drying:
    3.0 - 6.0 %
  • USP - Tartrate:
    Passes Test
  • USP - Assay (C₆H₅K₃O₇) (dried basis):
    99.0 - 100.5 %
  • FCC - Identification:
    Passes Test
  • FCC - Alkalinity:
    Passes Test
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Loss on Drying:
    3.0 - 6.0 %
  • FCC - Assay (C₆H₅K₃O₇) (dried basis):
    99.0 - 100.5 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together