Potassium citrate monohydrate 99.0-100.5% (dry basis), meets analytical specification of USP, Sigma-Aldrich®
: Merck
:6100-05-6
:
:HOC(COOK)(CH₂COOK)₂·H₂O
:MFCD00150442
:12,07785
:324.41 g/mol
:212-755-5
- Loss on Drying :3.0 - 6.0%
- Identity:Pass
- Alkalinity:Pass
- Assay (Dry Basis):99.0 - 100.5 %
- Tartrate:Pass
- Residual Solvents USP:Meets Requirements
Frequently Bought Together