
Disodium oxalate ≥99.5% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:62-76-0
:
Formula:Na₂C₂O₄
MDL Number:MFCD00012465
:134 g/mol
Merck Index:13,08723
Melting Pt:250…270 °C
Density:2.340 g/cm³ (20 °C)
EINECS:200-550-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity (IR):complying
- Assay:Min. 99.50 %
- Loss on drying (105°C):Max. 0.01 %
- Indifference:complying
- Water insoluble matter:Max. 0.005 %
- Iron (Fe):Max. 0.001 %
- Potassium (K):Max. 0.005 %
- Heavy metals (as Pb):Max. 0.002 %
- Chloride (Cl):Max. 0.002 %
- Sulfate (SO₄):Max. 0.002 %
- Ammonium (NH₄):Max. 0.002 %
- Reaction against H₂SO₄:complying
Specification Test Results
Frequently Bought Together