Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Disodium oxalate ≥99%, GPR RECTAPUR®
  27977.267
 :  
undefined
 
Disodium oxalate ≥99%, GPR RECTAPUR®
  27977.267
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 100 ppm

 

 

 :
 :134 g/mol
 :13,08723
 :250…270 °C
 :2.27 g/cm³ (20 °C)
 :200-550-3
 :GHS07
 :Ambient
 :62-76-0
 :Na₂C₂O₄
 :MFCD00012465