
Disodium oxalate ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:62-76-0
:
Formula:Na₂C₂O₄
MDL Number:MFCD00012465
:134 g/mol
Merck Index:14,8650
Melting Pt:250…270 °C (dec.)
Density:2.340
EINECS:200-550-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together