Malonic acid ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 141-82-2 Molecular Formula: C3H4O4 Formula Weight: 104.06 Flash Point: 157°(314°F) Melting Point: 132-136° dec. Beilstein Reference: 1751370 Merck Reference: 14,5710 MDL No.: MFCD00002707
Synonyms: 1,3-Propanedioic acid
- CAS Number:141-82-2
- Formula:C₃H₄O₄
- MDL Number:MFCD00002707
- Molecular Weight:104.06 g/mol
- Merck Index:14,5710
- Boiling Pt:387 °C (1013 hPa)
- Melting Pt:132…136 °C (dec.)
- Density:1.62
- Flash Pt:157 °C (314 °F)
- Storage Temperature:Ambient
- EINECS:205-503-0
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 141-82-2 Molecular Formula: C3H4O4 Formula Weight: 104.06 Flash Point: 157°(314°F) Melting Point: 132-136° dec. Beilstein Reference: 1751370 Merck Reference: 14,5710 MDL No.: MFCD00002707
Synonyms: 1,3-Propanedioic acid
- CAS Number:141-82-2
- Formula:C₃H₄O₄
- MDL Number:MFCD00002707
- Molecular Weight:104.06 g/mol
- Merck Index:14,5710
- Boiling Pt:387 °C (1013 hPa)
- Melting Pt:132…136 °C (dec.)
- Density:1.62
- Flash Pt:157 °C (314 °F)
- Storage Temperature:Ambient
- EINECS:205-503-0
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



