

Mercaptoacetic acid (thioglycolic acid) ≥98%, GPR RECTAPUR®
Catalog # 20495.265
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Assay (at filling):Min. 98 %
- Ignition residue (SO4):Max. 0.1 %
- Fe (Iron):Max. 5 ppm
Specification Test Results
About this item
:
:92.12 g/mol
Merck Index:13,09410
Boiling Pt:96 °C (1013 hPa)
Melting Pt:–16,5 °C
Density:1.33 g/cm³ (20 °C)
Flash Pt:130 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:200-677-4
UN:1940
ADR:8,II
:GHS06
:GHS05
CAS Number:68-11-1
Formula:HSCH₂COOH
MDL Number:MFCD00004876