
N-Lauroylsarcosine sodium salt, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:137-16-6
:
Formula:H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MDL Number:MFCD00042728
:293.38 g/mol
Merck Index:12,04379
Melting Pt:45…47 °C
Flash Pt:>150 °C
Storage Temperature:Ambient
EINECS:205-281-5
UN:2811
ADR:6.1,II
:GHS06
Frequently Bought Together