

Sebacoyl dichloride 92%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-19-3
:
Formula:C₁₀H₁₆Cl₂O₂
MDL Number:MFCD00000770
:239.14 g/mol
Boiling Pt:220 °C (75 mmHg)
Melting Pt:–2.5 °C
Density:1.121 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:203-843-4
UN:2922
ADR:8,II
:GHS06
:GHS05
Specification Test Results
- Appearance:Light yellow to golden to brown Liquid (May develop some turbidity or precipitate)
- Infrared spectrum:Conforms
- Refractive index:1.4670 to 1.4700 (20°C, 589 nm)
- GC:≥90 %
- Titration RCOCl:≥90 %
Specification Test Results
Frequently Bought Together