Diethyl adipate for synthesis, Sigma-Aldrich®
: Merck
:141-28-6
:
:C₁₀H₁₈O₄
:MFCD00009215
:202.25 g/mol
:13,00163
:∼251 °C (1013 hPa)
:–19.8 °C
:1.01 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:>100 °C
:Ambient
:205-477-0
- Solubility:- 0.1 g/l (20 °C)
- Melting Point:-19.8 °C
- Molar Mass:202.25 g/mol
- Boiling Point:245 °C (1013 hPa)
- Flash Point:100 °C
- Density:1.01 g/cm3 (20 °C)
Frequently Bought Together