Bis(2-ethylhexyl)sebacate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:122-62-3
:
Formula:C₂₆H₅₀O₄
MDL Number:MFCD00009497
:426.68 g/mol
Merck Index:13,01251
Boiling Pt:435 °C (1013 hPa)
Melting Pt:–55 °C
Density:0.914 g/cm³ (20 °C)
Flash Pt:213 °C
Storage Temperature:Ambient
EINECS:204-558-8
Frequently Bought Together