Order Entry
Czech Republic
ContactUsLinkComponent
 
Bis(2-ethylhexyl)sebacate for synthesis, Sigma-Aldrich®
  8.14576.1000
 :  Merck
 :  
undefined
Bis(2-ethylhexyl)sebacate for synthesis, Sigma-Aldrich®
  8.14576.1000
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Solubility:
     - 0.1 g/l (20 °C)
  • Melting Point:
    -55 °C
  • Molar Mass:
    426.68 g/mol
  • Boiling Point:
    250 °C (1013 hPa)
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    200 °C
  • Density:
    0.91 g/cm3 (20 °C)

 

 

 :
 :426.68 g/mol
 :13,01251
 :435 °C (1013 hPa)
 :–55 °C
 :0.914 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :213 °C
 :Ambient
 :204-558-8
 :122-62-3
 :C₂₆H₅₀O₄
 :MFCD00009497