

Isopropyl formate, Sigma-Aldrich®
Catalog # SIAL476455-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:88.11 g/mol
Boiling Pt:67…68 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.8826 g/cm³ (0 °C)
EINECS:210-901-2
UN:1281
ADR:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
CAS Number:625-55-8
Formula:C₄H₈O₂
MDL Number:MFCD00014127