

Urea 98.0-102.0% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:57-13-6
:
Formula:NH₂CONH₂
MDL Number:MFCD00008022
Merck Index:13,09935
Boiling Pt:197 °C (1013 hPa)
Melting Pt:132.7 °C
Density:1.32 g/ml (20 °C)
:60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
REACH:01-2119463277-33
:GHS07
:GHS08
Specification Test Results
- Meets U.S.P Requirements:Passes Test
- GMP Manufactured Product:Passes Test
- Meets B.P. Chemical Specifications:98.0 - 102.0%
- Meets E.P. Chemical Specifications:≤0.1%
- Meets J.P. Chemical Specifications:≤0.04%
- CAUTION: For Manufacturing, processing or repackaging:≤0.1%
- Bulk Pharmaceutical Chemical:≤0.1%
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Assay (NH₂CONH₂):98.0 - 102.0 %
- USP - Residue on Ignition:≤0.1 %
- USP - Alcohol Insoluble Matter:≤0.04 %
- USP - Organic impurities: Urea related compound A:≤0.1 %
- USP - Organic impurities: Any individual unspecified impurity:≤0.1 %
- USP - Organic impurities: Total impurities:≤2.0 %
- USP - Endotoxin Concentration (EU/g):≤2.5
- EP/BP - Assay (NH₂CONH₂) (dried basis):98.5 - 101.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Alkalinity:Passes Test
- EP/BP - Biuret:≤0.1 %
- EP/BP - Ammonium (NH₄):≤500 ppm
- EP/BP - Loss on Drying at 105°C:≤1.0 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (NH₂CONH₂):99.0 - 101.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Chloride (Cl):≤0.007 %
- JP - Sulfate (SO₄):≤0.010 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Alcohol Insoluble Matter:≤0.040 %
- JP - Residue on Ignition:≤0.10 %
- JP - Melting Point:132.5 - 134.5 °C
Specification Test Results
Related Information
Frequently Bought Together