Propionamide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-05-0
:
Formula:C₃H₇NO
MDL Number:MFCD00008039
:73.09 g/mol
Merck Index:12,08009
Boiling Pt:213 °C (760 mmHg)
Melting Pt:79…83 °C
Density:1,1 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:201-172-1
Specification Test Results
- Appearance:Colorless to white to light yellow Powder or crystals or flakes and/or chunks
- Infrared spectrum:Conforms
- Melting point:77 to 83 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together