Formamide, Sigma-Aldrich®
Catalog # SIALF9037-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
Merck Index:13,04261
Boiling Pt:210 °C (1013 hPa)
Melting Pt:2 °C
Density:1.1334 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:79 °C
:45.04 g/mol
Storage Temperature:Refrigerator
EINECS:200-842-0
:GHS08
CAS Number:75-12-7
Formula:HCONH₂
MDL Number:MFCD00007941