Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:75-12-7
:
Formula:HCONH₂
MDL Number:MFCD00007941
Merck Index:13,04261
Boiling Pt:210 °C (1013 hPa)
Melting Pt:2 °C
Density:1.1334 g/cm³ (20 °C)
Flash Pt:79 °C
:45.04 g/mol
EINECS:200-842-0
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Assay (on anhydrous substance):Min. 99.0 %
- Heavy metals (as Pb):Max. 1 ppm
- Ignition residue (SO4):Max. 50 ppm
- Water:Max. 0.2 %
- Cl (Chloride):Max. 1 ppm
- Fe (Iron):Max. 1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
24311_FORMAMIDE_BSE_REV0.pdf
BSE/TSE Certificates
spec_VWRC24311.291 spec_VWRC24311.320
Product specification
Frequently Bought Together