Orders LinkContactUsLinkComponent
Formamide ≥99.5% ACS
Formamide ≥99.5% ACS
Formamide ≥99.5% ACS
Formamide ≥99.5% ACS
Purity: 99.5+% CAS No.: 75-12-7 Molecular Formula: CH3NO Formula Weight: 45.04 Flash Point: 154°(310°F) Physical Form: Liquid Density: 1.134 Refractive Index: 1.4475 Sensitivity: Hygroscopic Appearance: Colourless Odour: Ammonia-like Melting Point: 2° Boiling Point: 210° Beilstein Reference: 505995 Merck Reference: 14,4237 MDL No.: MFCD00007941
 :  Carbamaldehyde
  •  :75-12-7
  •  :HCONH₂
  •  :MFCD00007941
  •  :45.04 g/mol
  •  :14,4237
  •  :210 °C
  •  :2 °C
  •  :1.134
  •  :154 °C (310 °F)
  •  :Ambient
  •  :200-842-0
Product Family Options
  • Pack type
    Pk
  • 4
    1 * 4 KS
    1 * 4 KS - 16 640,00 Kč
    16 640,00 Kč
    1 * 4 KS
       16 640,00 Kč
  • Glass bottle for solids
    250 g
    1 * 250 g
    1 * 250 g - 2 570,00 Kč
    2 570,00 Kč
    1 * 250 g
       2 570,00 Kč
  • Glass bottle for solids
    1 kg
    1 * 1 kg
    1 * 1 kg - 4 650,00 Kč
    4 650,00 Kč
    1 * 1 kg
       4 650,00 Kč