Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

N,N-Dimethylformamide ≥99%

Health
Flammable
Irritant
Marketsource
N,N-Dimethylformamide ≥99%
Marketsource
Certificates
N,N-Dimethylformamide ≥99%
N,N-Dimethylformamide ≥99%

4 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 68-12-2 Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 Flash Point: 57°(135°F) Refractive Index: 1.4310 Odour: Amine-like pH: 7 Melting Point: -61° Boiling Point: 153° Beilstein Reference: 605365 Merck Reference: 14,3243 UN No.: 2265 MDL No.: MFCD00003284

Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:14,3243
  • Boiling Pt:153 °C
  • Melting Pt:–61 °C
  • Density:0.944
  • Flash Pt:57 °C (135 °F)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32
Product Family Options

Product Information