N,N-Dimethylformamide 99.8% for molecular biology DNase-, RNase-, protease-free
:68-12-2
:
:HCON(CH₃)₂
:MFCD00003284
:73.09 g/mol
:13,03269
:153 °C (760 mmHg)
:–61 °C
:0.945 g/cm³
:58 °C
:Ambient
:200-679-5
:2265
:3,III
:01-2119475605-32
:GHS07
:GHS08
:GHS02
- Appearance:Clear liquid
- Water:≤0.05 % (Coulometric)
- GC:≥99.8 %
- UV:(undiluted)
- Color scale:≤5 APHA
- Residue after evaporation:≤0.001 %
- Dnase,Rnase,Protease activity:none detected
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Free amine:≤0.001 % (Kaiser)
- Acidity (CH3COOH):≤0.001 %
Frequently Bought Together